Learn More
Medchemexpress LLC Thiourea, N-[3,5-bis(trifluoromethyl)phenyl]-N'-(phenylmethyl)- | 13571-44-3 | 99.9% | 378.34 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY147291
Encompass_Preferred
VPC-70063 is a potent Myc-Max inhibitor with an IC50 value of 8.9 μM for Myc-Max transcriptional activity inhibition. It reduces UBE2C promotor activity and AR-V7 levels, induces PARP cleavage, apoptosis, and blocks Myc-Max interactions with DNA. VPC-70063 can be used for researching anticancer.
  • Potent Myc-Max inhibitor
  • Reduces UBE2C promotor activity and AR-V7 levels
  • Induces PARP cleavage and apoptosis
  • Blocks Myc-Max interactions with DNA
  • Suitable for anticancer research
Catalog No. 50-003-63705