Learn More
Medchemexpress LLC Thiq | 312637-48-2 | MFCD09970598 | 98.5% | 589.17 | C33H41ClN6O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1062410MG
Encompass_Preferred
THIQ is a selective melanocortin-4 receptor (MC4R) agonist used in research to probe MC4R signaling and receptor trafficking. It functions as a pharmacoperone that can rescue cell-surface expression and signaling of certain intracellularly retained MC4R mutants and has been employed in rodent studies of erectile activity.
  • Selective MC4R agonist with high affinity and potency (hMC4R IC50=1.2 nM; EC50=2.1 nM; rMC4R IC50=0.6 nM; EC50=2.9 nM).
  • Acts as a pharmacoperone to restore receptor cell-surface expression.
  • High reported purity and supplied as a solid, white to off-white material.
  • Stable under recommended storage (powder: -20°C for 3 years; 4°C for 2 years).
  • Suitable for in vitro and in vivo studies of melanocortin receptor biology.
Catalog No. 50-002-42897