Learn More
New Era Pump Systems Inc Third+ Syringe Heating Pad 120Vac 33W, 54cm cord, 9cm x 7.5cm (works w/ HEATER-KIT-1LG w/ a HEATER-PAD2-1LG)
Supplier:  New Era Pump Systems Inc HEATERPAD31LG
Encompass_Preferred
Third+ Syringe Heating Pad 120Vac 33W, 54cm cord, 9cm x 7.5cm (works w/ HEATER-KIT-1LG w/ a HEATER-PAD2-1LG)
Catalog No. NC2258317