Learn More
Bachem N-Me-DL-Phe-OMe · HCl
SDP
Supplier:  Bachem 4027307.0005
Encompass_Preferred
N-Me-DL-Phe-OMe · HCl, 16975-45-4, C11H15NO2 · HCl, Mr 229.71, 5g. This building block has been used in the synthesis of 1,4-benzodiazepine-2,5-diones.
Catalog No. 50-194-7257