Learn More
Bachem (DL-Isoser1)-TRAP-6 Trifluoroacetate
SDP
Supplier:  Bachem 4025390.0001
Encompass_Preferred
(DL-Isoser1)-TRAP-6 Trifluoroacetate, H-DL-Isoser-Phe-Leu-Leu-Arg-Asn-OH, 150242-29-8, C34H56N10O9, Mr 748.88, 1mg. (DL-Isoser1)-Thrombin Receptor Activator Peptide 6, iso-TRAP, (DL-Isoser1)-PAR-1 (1-6) (human), (DL-Isoser1)-Proteinase Activated Receptor 1 (1-6) (human), (DL-Isoser1)-Thrombin Receptor 1 (1-6) (human). This racemic isoserine analog of TRAP-6 (SFLLRN) has only 15-20% of the platelet aggregating activity of SFLLRN. In compensation iso-SFLLRN significantly resists degradation by aminopeptidase M and is much more stable in plasma and serum.
Catalog No. 50-194-6790