Learn More
Perkin Elmer US LLC Diffuse Reflectance Reference Materials Standards - Calibrated, Qty. 8
Supplier:  Perkin Elmer US LLC PELA9012
Encompass_Preferred
PerkinElmer's Diffuse Color and Neutral Reference Material sets are used to establish the photometric accuracy and linearity of the LAMBDA 850+ and 1050+ spectrometers. These reference materials are supplied with calibration data from 250 to 2500 nm, reported every 50 nm. Sets also included a disk with data for each 1 nm. Reflectance calibration is traceable to NIST and NPL standards.
Catalog No. 50-201-6475
Only null left
Add to Cart
Request a Quote