Learn More
Medchemexpress LLC HY-101183 1mg Medchemexpress, THK5351 CAS:1707147-26-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1011831MG
Encompass_Preferred
Medchemexpress, HY-101183 1mg THK5351 CAS:1707147-26-9 THK5351 can be radiolabeled and used as a radiotracer for in vivo imaging of tau pathology in the brain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2060