Learn More
ERA WATERS CORP THMS QC STD
SDP
Supplier:  ERA WATERS CORP 702
Encompass
One 2 mL flame-sealed ampule yields in excess of 200 mL after dilution. Contains all of the analytes below at 5-50 ug/L.
Catalog No. NC9738460