Learn More
Thomas Scientific Foil Squares, 6" X 6", 1 Mil Thick
SDP
Supplier:  Thomas Scientific 1086J26/DEL
Encompass_Preferred
Pre-cut aluminum foil for convenience For weighing, capping, glassware covers, lids, etc. Pre-cut aluminum foil squares are interleaved with paper and furnished in a 5 lb. (2.3 kg) package. Package quantities are approximate.
Catalog No. NC9582171