Learn More
Thomas Scientific Spectro Cell 334-2500 nm
SDP
Supplier:  Thomas Scientific 8491D03
Encompass_Preferred
Minimum working capacity as shown in listings Cell bodies are of fused-wall construction. Matched sets are supplied in cushioned storage box. Includes Teflon cover. 8491M18 cells require 8494J20 adapter to accommodate cells in standard cell carrier.
Catalog No. NC9197440