Learn More
Thomas Scientific Sample Wrapper, Ogg, Black
SDP
Supplier:  Thomas Scientific 6514F70/DEL
Encompass_Preferred
6514F35 Ogg Flask of borosilicate glass, with Standard Taper 35/25 socket mouth; without stopper or sample carrier. 1233S49 Platinum Ogg Sample Carrier has a perforated aluminum sheet. It features a 13 mm deep basket on lower end that takes the paper-wrapped sample. 6514F40 Stopper fits the 1,000 mL flask; glass hook tip on lower end; platinum sample carrier not included. 6514F70 Black Paper Sample Wrappers absorb heat quickly for infrared ignition; die-cut from unsized paper; sample area 32 x 30 mm, with 8 x 38 mm extending strip as fuse for ignition.
Catalog No. NC9440438
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product