Learn More
Thomas Scientific Silicone Fluid | 16-oz
SDP
Supplier:  Thomas Scientific 6428R15
Encompass_Preferred
This clear, colorless, oily liquid has a nominal viscosity of 50 centistokes at 25°C, with a viscosity/ temperature coefficient of 0.59. The boiling point is over 360°C with a flash point above 535°F. The specific gravity is 0.963; the refractive index, 1.402. Surface tension is 20.8 dynes/cm, all at 25°C. Fluid has a maximum volatility, with 0.5% weight loss over 24 hours at 150°C.
Catalog No. NC9495670