Learn More
Thomas Scientific Silicone Fluid 1/2 Gl
SDP
Supplier:  Thomas Scientific 6428R25
Encompass_Preferred
Silicone bath fluid Superior heat transfer medium in melting point baths, etc. High thermal and chemical stability This clear, colorless, oily liquid has a nominal viscosity of 50 centistokes at 25ÝC, with a viscosity/ temperature coefficient of 0.59. The boiling point is over 360ÝC with a flash point above 535ÝF. The specific gravity is 0.963; the refractive index, 1.402. Surface tension is 20.8 dynes/cm, all at 25ÝC. Fluid has a maximum volatility, with 0.5% weight loss over 24 hours at 150ÝC.
Catalog No. NC9704805