Learn More
Thomas Scientific Blade Handle
SDP
Supplier:  Thomas Scientific 6727C25
Encompass_Preferred
Used frequently for preparation of thin, uniform sections of fresh tissue as necessary for metabolic studies of tissue in vitro Sections average 0.5 mm in thickness Consists of three flat plates of carefully machined and polished transparent methacrylate, 13 mm thick, one extended to form a handle and a thin carbon steel blade with 102 mm cutting edge; not autoclavable Nickel-plated brass, detachable handle with wing nut for fastening blade Parts separate easily for cleaning Tissue Slicer (part no. 6727C10) comes with a 25 pk of blades (6727C18), blade handle (6727C25), and 3 trans. methacrylate parts (6727P02) Circular platform, 31 mm diameter x 12 mm high, on base plate; fits into circular opening in middle plate. With 10 carbon steel blades, 117 mm long. Other Applications: these blades are also used in the polymer clay industry for thin slicing of clay materials, with no pulling or tearing of material. It also produces very thin sections when needed.  NOTE: these blades are ex
Catalog No. NC9992568