Learn More
TARGETMOL CHEMICALS INC THONZONIUM BROMIDE 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T218850MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Thonzonium bromide is a mono-cationic surfactant that is structurally similar to Farnesol as an antimicrobial agent. It inhibits proton transport in a dose-dependent manner (EC50=69 uM). It inhibits RANKL-induced osteoclast formation and bone resorption in vitro and prevents LPS-induced bone loss in vivo. purity: 99%
Catalog No. 50-258-0992