Learn More
Medchemexpress LLC Thp-peg1-alcohol | 2162-31-4 | MFCD00185588 | 99.5% | 146.18 | C7H14O3 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW038771
Encompass_Preferred
THP-PEG1-alcohol is a THP-protected, PEG-based linker bearing a terminal alcohol group used as a building block in the synthesis of PROTACs and other bifunctional molecules. The short PEG spacer provides compact linker length with improved solubility, while the THP group protects the alcohol during multi-step syntheses. Intended for research use only.
  • Thp-protected alcohol functionality enables selective deprotection under mild acidic conditions.
  • Short PEG spacer (PEG1) gives a compact, flexible linker with good solubility.
  • High purity (99.5%) suitable for research-grade synthesis.
  • Supplied with certificate of analysis and safety data sheet for traceability.
  • Available in gram-scale quantities for synthetic workflows.
Catalog No. 50-003-03558