Learn More
Medchemexpress LLC Thp-peg1-alcohol | 2162-31-4 | MFCD00185588 | 99.5% | 146.18 g/mol | C7H14O3 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0387715G
Encompass_Preferred
THP-PEG1-alcohol is a THP-protected PEG1 alcohol used as a short PEG linker building block for the synthesis of PROTACs and related conjugates. The tetrahydropyranyl (THP) protecting group preserves the terminal alcohol during multi-step synthesis and can be removed under mild acidic conditions to reveal a reactive hydroxyl for further functionalization.
  • THP-protected terminal alcohol enabling selective deprotection under mild acidic conditions.
  • Short PEG spacer (one ethylene glycol unit) for minimal linker length and low steric bulk.
  • High purity (99.53%) suitable for synthetic and research applications.
  • Chemical formula C7H14O3 and molecular weight 146.18 g/mol.
  • Liquid form, colorless to light yellow, compatible with standard organic solvents.
Catalog No. 50-003-00388