Learn More
Medchemexpress LLC Thp-peg4-pyrrolidine(n-boc)-ch2oh | 2378261-80-2 | 95.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13082050MG
Encompass_Preferred
THP-PEG4-Pyrrolidine(N-Boc)-CH2OH is a PEG-based PROTAC linker that can be used in the synthesis of PROTAC K-Ras Degrader-1 (HY-129523). It is for research use only. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTAC K-Ras degrader-1
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-40973