Learn More
Medchemexpress LLC Thp-peg6-oh | 42607-87-4 | 366.45 | C17H34O8 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY126918250MG
Encompass_Preferred
THP-PEG6-OH is a PEG-based linker used for constructing proteolysis targeting chimeras (PROTACs) and as a non-cleavable 3 unit PEG linker for antibody-drug conjugates (ADCs). It is supplied as a solid for research use in medicinal chemistry and bioconjugation workflows.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Non-cleavable 3 unit PEG ADC linker
  • Suitable for ADC synthesis and bioconjugation
  • Available in multiple pack sizes including 250 mg
Catalog No. 50-002-04204