Learn More
ATCC THP-1; Acute Monocytic Leukemia; Human (Homo sapiens)
Supplier:  ATCC TIB202
Encompass
THP-1 is a monocyte isolated from peripheral blood from an acute monocytic leukemia patient. This cell line can be used in immune system disorder research, immunology research, and toxicology research. Note: These are suspension cells and floating cells are viable.
Catalog No. 50-238-2192
 Restricted Item