Learn More
Cyclodextrin Technologies Development Hydroxypropyl BCD, 31g
Supplier:  Cyclodextrin Technologies Development THPBP31G
Encompass
Beta Cyclodextrin derivative for enhanced solubility >50g/100ml - meets USP/EP standards. Pharmaceutical Grade
Catalog No. NC0137397