Learn More
Medchemexpress LLC THR-β agonist 1 | 2648403-17-0 | 98.7% | C17H14Cl2FN5O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14244110MM/1ML
Encompass_Preferred
THR-β Agonist 1 is a potent and selective agonist of THRβ. Thyroid hormone receptor mediates the physiological activity of thyroid hormones, which plays a key role in normal growth and development of the body and in maintaining metabolic balance. It has potential for the research of thyroid hormone receptor-related diseases.
  • Potent and selective agonist of THRβ
  • Mediates the physiological activity of thyroid hormones
  • Plays a key role in normal growth and development of the body
  • Maintains metabolic balance
  • Potential for research of thyroid hormone receptor-related diseases
Catalog No. 50-003-55580