Learn More
Sino Biological Peptide Substrates: Thr-phosphopeptide
Supplier:  Sino Biological T6958
Encompass_Preferred
The Thr-phosphopeptide sequence (KRT(p)IRR) is derived from human NR2F6 (amino acids 79-84) and is suitable as a substrate for PP2A, PP2B, PP2C and PP1.
Catalog No. 50-256-7584