Learn More
Grainger Thread Mill 1 2 -11.5 Carbide
Supplier:  Grainger 16R660
Encompass_Preferred
The Micro 100 16R660 is a 1/4" diameter, 3/4" long, 4 flute carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree threads from 1-72 TPI in steel, stainless steel, cast iron, aluminum, and other materials. The TiAlN coating provides heat and wear resistance. This thread mill is made in the USA by Micro 100 and is well-suited for production threading applications.
Catalog No. 50-276-7696