Learn More
Grainger Thread Mill 1/4 3/8 -18 Carbide
Supplier:  Grainger 16R670
Encompass_Preferred
The Grainger 16R670 is a Westinghouse 15 Amp, 125 Volt, 3-Wire Grounding Plug with Straight Blades and Side Wire Holes. It has a NEMA 5-15P configuration with a black, white and green wire connection and is UL Listed. The plug is compatible with 14 AWG wire and has side wire holes for easy installation. It can be used on lamps, small appliances, power tools and more. The plug measures 1-3/4" long x 1-1/8" wide x 1-1/4" high. This general use plug provides a safe and secure electrical connection.
Catalog No. 50-276-8469