Learn More
Grainger Thread Mill 5/16 -24 Carbide
Supplier:  Grainger 16R697
Encompass_Preferred
The THD Mill Single Int Ext 5/16-16 48 TPI is a high-speed steel milling cutter with a single internal and external tooth design. It has a 5/16" shank diameter, an overall length of 1-1/2", and 48 teeth per inch. This mill is made of M2 high-speed steel for wear resistance and has TiN coating for increased hardness and lubricity. It can be used for slotting, side milling and profiling in both ferrous and non-ferrous materials. The geometry allows for smooth, clean cuts. This is a quality milling tool well-suited for machine shops.
Catalog No. 50-276-5712