Learn More
Chemglass Life Sciences Chem-Thread, Size 9, 13mm Tube OD, Accepts 7-9mm OD Tubing, #110 O-ring
SDP
Supplier:  Chemglass Life Sciences CG35002
Encompass_Preferred
Chem-Thread, Size 9, 13mm Tube OD, Accepts 7-9mm OD Tubing, #110 O-ring. Consists of an externally threaded glass tube, Viton o-ring and Celcon compression cap. Tube O.D. listed is the shank O.D. of the connector.
Catalog No. 50-873-752