Learn More
MSC Balax Thread Plug Gage, 3-56 Thread, 2B Class, Double End, 1 ea
Supplier:  MSC 05363577
Encompass
This thread plug gage is designed for precise inspection of internal threads, featuring a double end configuration with both Go and No Go ends to ensure accuracy in thread production. Constructed from high-speed tool steel with a bright finish, this tool provides reliable performance for checking Class 2B fit threads.
  • Manufactured from high-speed tool steel for durability
  • Includes both Go and No Go ends for comprehensive thread inspection
  • Designed for 3-56 thread size with 2B class of fit
  • Hardness rating of 63 to 65 Rockwell
  • Compatible with #000 Taperlock handles
  • Country of origin is United States
Catalog No. 50-005-16888