Learn More
MSC Balax Thread Plug Gage, 12-28 Thread, 3B Class, Double End, 1 ea
Supplier:  MSC 05369319
Encompass
This thread plug gage is designed for precision inspection of internal threads, ensuring they meet specified tolerances. It features a double-end construction with Go and No Go members for efficient verification of threaded holes in metalworking and inspection applications. Engineered from high-speed tool steel for durability and accurate measurement, this tool is built to withstand repeated use in demanding industrial environments.
  • Designed with 12-28 thread size and 3B class fit for precise thread inspection
  • Constructed from high-speed tool steel with a hardness of 63 to 65 Rockwell
  • Features a double-end configuration with Go and No Go members
  • Equipped with a #0 taperlock handle
  • Finished with a bright coating
Catalog No. 50-005-11180