Learn More
MSC GF Gage Thread Ring Gage, 1-1/4-7 Thread, Class 2A, No Go, 1 ea
Supplier:  MSC 55323638
Encompass
This threaded ring gage is designed for precision inspection of external threads, specifically to verify that dimensions fall within the required tolerance range. It is configured as a no-go gage to ensure that the produced thread does not exceed specified limits, making it a critical tool for quality control in metalworking and machining applications.
  • Features a 1-1/4-7 thread size
  • Class 2A classification for standard thread fit accuracy
  • Designed as a single, no-go threaded ring gage
  • Suitable for industrial thread inspection processes
  • Manufactured in the United States
Catalog No. 50-005-47858