Learn More
Cole-Parmer Idex Threaded Luer Adapter, Natural/Black PEEK, 0.050" Bore, Male Slip Luer x Female 10-32 Coned; 1/EA
Supplier:  Cole-Parmer 0202357
Encompass
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.Manufactured from inert polymersMale Slip Luer x Female 10-32 Coned0.050" thru-hole diameterManufactured from PEEK
Catalog No. NC3737991