Learn More
Grainger Threaded Shank Drill 1/8 HSS
Supplier:  Grainger 16W695
Encompass_Preferred
The Grainger 1/16" Threaded Shank Drill Bit is a high-speed steel bit with a 1/16" diameter and 1/4" shank. It has a straight flute design and a 118 degree point angle for drilling into metal, wood, and plastic. The bit features a threaded shank that allows it to be securely fastened into a drill chuck. Made in the USA, this drill bit is designed for precision drilling in light-duty applications. With proper use, it provides accurate holes with minimal burring.
Catalog No. 50-276-4227