Learn More
Grainger Threaded Shank Drill #13 Cobalt
Supplier:  Grainger 16W771
Encompass_Preferred
The Grainger 13/16" Threaded Shank Drill Bit is made of high-speed steel and has a black oxide finish for durability. It features a 135 degree split point tip for faster penetration and reduced walking. The bit has a 1/2" hex shank to fit power drills. It can drill through metal, wood, and plastic. Overall length is 5-1/2". Made in the USA.
Catalog No. 50-276-4824