Learn More
Perkin Elmer US LLC Threaded Spray Chamber Drain Line with Santoprene Tubing for NexION 2000
Supplier:  Perkin Elmer US LLC N8152391
Encompass_Preferred
The drain line kit includes a 1/4-28 spray chamber fitting with attached 32 cm line, as well as additional 0.75 mm I.D. Santoprene MPP tubing (double grey marked).
Catalog No. 50-211-2139