Learn More
Grainger Threading Tool 1/8 Carbide
Supplier:  Grainger 16T082
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with an 8mm shank diameter and 16mm maximum cutting diameter. It has a 60 degree threading profile and can cut metric, UN, and UNF threads. The overall tool length is 3.5 inches. This single-point threading tool allows both external and internal threading operations. It features TiN coating for increased tool life and lubricity. The Micro 100 threading tool is made in the USA.
Catalog No. 50-276-4087