Learn More
Grainger Threading Tool 3/16 Carbide
Supplier:  Grainger 16T311
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree threading point and can cut unified coarse right hand threads. The overall length is 3-1/2" and it has a TiN coating for increased tool life. This threading tool is good for production threading applications in common steels, cast iron, alloy steels, stainless steels, and exotic metals. It has a screw lock style holder for secure clamping.
Catalog No. 50-276-5996