Learn More
Grainger Threading Tool 3/16 Carbide
Supplier:  Grainger 16T302
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree threading point and can cut unified coarse right hand threads. The overall length is 3-1/2" and it weighs 0.19 lbs. This single point threading tool allows quick threading starts and smooth surface finishes. It is made in the USA.
Catalog No. 50-276-6567