Learn More
Grainger Threading Tool 3/16 Carbide
Supplier:  Grainger 16T265
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 60 degree included angle. It has a 0.040 inch minimum minor diameter and can thread up to 5/8 inch depth of thread. The overall tool length is 3 inches. This single point threading tool allows both right hand and left hand threading. It has TiN coating for wear resistance and lubricity. The Micro 100 threading tool is made in the USA.
Catalog No. 50-276-7204