Learn More
Grainger Threading Tool 3/8 Carbide
Supplier:  Grainger 16T127
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree included angle and can cut unified thread forms. The overall length is 5" and it weighs 0.19 lbs. This single point threading tool allows quick threading passes with precision and accuracy. It is made in the USA out of M2 high speed steel for strength and durability. The Micro 100 threading tool can be used on lathes, screw machines, and CNC machines. It is ideal for both production threading and general purpose workshop threading applications.
Catalog No. 50-276-4893