Learn More
Grainger Threading Tool 3/8 Carbide
Supplier:  Grainger 16T377
Encompass_Preferred
The Micro 100 3/8" shank threading tool is a high speed steel insert style threading tool with a TiN coating for wear resistance. It has a 60 degree threading insert that can cut unified, metric, ACME, buttress, and worm threads. The tool has an overall length of 4" and a shank diameter of 3/8". It is designed for use on CNC lathes, screw machines, and manual lathes. The tool comes with one DCMT 21.51 threading insert and can cut thread sizes from #4-40 to 5/8"-11.
Catalog No. 50-276-8597