Learn More
Grainger Threading Tool 5/16 Carbide
Supplier:  Grainger 16T361
Encompass_Preferred
The Grainger 5/16"-16-48 single profile thread mill is a high-speed steel thread milling cutter with a 5/16"-16 thread profile and a 48mm overall length. It has a TiN coating for wear resistance and can be used on materials like steel, stainless steel, cast iron, aluminum and more. The cutter has a 1/2" shank diameter and comes with a Rc hardness of 63-65 to handle interrupted cuts. It is designed for right hand threads.
Catalog No. 50-276-7112