Learn More
Grainger Threading Tool 5/16 Carbide
Supplier:  Grainger 16T119
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with a 5/16" shank diameter and 16 TPI. It has a 60 degree threading point and can cut unified coarse right-hand threads. The overall length is 3-1/2" and it has a TiN coating for increased tool life. This single-point threading tool allows quick threading passes on lathes, screw machines, and CNC lathes. It is made in the USA.
Catalog No. 50-276-6085