Learn More
Grainger Threading Tool 6.00mm Carbide
Supplier:  Grainger 16T198
Encompass_Preferred
The Grainger Approved 6.00mm Carbide Threading Tool is a high-speed steel threading tool with a 6mm cutting diameter. It has a 55 degree included angle and features a TiN coating for increased hardness and lubricity. The tool has an overall length of 4" and a shank diameter of 3/8". It is designed for right-hand threads and internal threading applications. The carbide construction provides strength and durability for cutting tough materials like stainless steel, alloy steels, cast iron, and hardened steels up to Rc50.
Catalog No. 50-276-8624