Learn More
Cole-Parmer Traceable Three-Scale Dial Barometer with Calibration; mbar/"Hg/mm Hg
Supplier:  Cole-Parmer 9976050
Encompass_Preferred
Three-Scale dial barometer measures atmospheric pressure in millibar (mbar), inches Hg, and mm Hg. The barometer is housed in corrosion-proof nickel-chrome case. Set the adjustable pointer to measure changes in pressure. Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Catalog No. NC1318456