Learn More
CPC Scientific azidoacetyl alanine-Lys-(5FAM)-Gly-Ala-Leu-Val-Pro-Arg-Gly-Ser-Ala-Gly-Lys(CPQ2)-NH2 (ammonium salt) 5MG
SDP
Supplier:  CPC Scientific THRO001B
Encompass
SEQUENCE: azidoacetyl alanine-Lys-(5FAM)-Gly-Ala-Leu-Val-Pro-Arg-Gly-Ser-Ala-Gly-Lys(CPQ2)-NH2 (ammonium salt)ONE-LETTER SEQUENCE: azidoacetyl alanine-K(5FAM)GALVPRGSAGK(CPQ2)-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT: 2143STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: HematologyCITATIONS:Zhu, Shu, et al. Blood 126.12 (2015): 1494-1502.SKU(s): THRO-001A, THRO-001B
Catalog No. 50-211-0562