Learn More
Cayman Chemical Thromboxn B2-d4 25ug
SDP
Supplier:  Cayman Chemical 31903025 UG
Encompass_Preferred
An internal standard for the quantification of TXB2 by GC- or LC-MS
Catalog No. 50-294-8330