Learn More
Cayman Chemical THUNDERHumn IL1 TRFRET 2400Wel
SDP
Supplier:  Cayman Chemical 5002302400 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-1β (IL-1F2) in cell culture supernatants
Catalog No. 50-289-5254