Learn More
Cayman Chemical THUNDERHumn IL12IL23 p4 96
SDP
Supplier:  Cayman Chemical 50023696 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-12/IL-23 p40 (IL-12B; NKFS; CLMF) in cell culture supernatants
Catalog No. 50-289-6390