Learn More
Cayman Chemical THUNDERHumn IL18 TRFRET 96
SDP
Supplier:  Cayman Chemical 50030496 WELL
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL‑18 (IL-1F4) in cell culture supernatants
Catalog No. 50-289-6195