Learn More
Cayman Chemical THUNDERPhospho-eIF2aS51 96Wel
SDP
Supplier:  Cayman Chemical 50020396 WELL
Encompass_Preferred
A sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-EIF2α (S51) in cell lysates
Catalog No. 50-289-6966